missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelial cell marker Antibody (LH39) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2690-0.025mg
This item is not returnable.
View return policy
Description
Epithelial cell marker Monoclonal antibody specifically detects Epithelial cell marker in Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| Epithelial cell marker | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LH39 | |
| Unconjugated | |
| PBS | |
| Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed in Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions. | |
| 0.025 mg | |
| Cancer, Extracellular Matrix | |
| Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction