missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epithelial cell marker Antibody (LH39) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 453.00
Specifications
| Antigen | Epithelial cell marker |
|---|---|
| Clone | LH39 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18578462
|
Novus Biologicals
NB100-2690-0.025mg |
0.025 mg |
€ 190.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18524293
|
Novus Biologicals
NB100-2690 |
0.1 mg |
€ 453.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epithelial cell marker Monoclonal antibody specifically detects Epithelial cell marker in Primate samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| Epithelial cell marker | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Extracellular Matrix | |
| PBS | |
| IgG1 | |
| Protein A purified |
| LH39 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Primate | |
| Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed in Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title