Learn More
Ethanesulfonic acid sodium salt, 98%, may cont. ca 2% water
Product Code. 11474857
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474857 5 g 5g
11464857 1 g 1g
11484857 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11474857 Supplier Thermo Scientific Alfa Aesar Supplier No. B21824.06
Request Bulk or Custom Format
CAS: 5324-47-0 | C2H5NaO3S | 132.109 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5324-47-0
Molecular Formula C2H5NaO3S
Molecular Weight (g/mol) 132.109
MDL Number MFCD00066498
InChI Key KQFAFFYKLIBKDE-UHFFFAOYSA-M
Synonym sodium ethanesulfonate, sodium ethanesulphonate, ethanesulfonic acid sodium salt, ethanesulfonic acid, sodium salt, ethane sulfonic acid sodium salt, sodiumethanesulfonate, sodium ethylsulfonate, sodium esylate, sodium ethane sulfonate, ethylsulfonic acid sodium salt
PubChem CID 4192170
IUPAC Name sodium;ethanesulfonate
SMILES CCS(=O)(=O)[O-].[Na+]
Specifications
Melting Point ∼270°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 132.11
Percent Purity 98%
Assay may cont. ≈2% water
Chemical Name or Material Ethanesulfonic acid sodium salt