Learn More
Ansell ETHIPARAT Ethylene Vinyl Acetate (EVA) Gloves, Non-Sterile
Product Code. p-7244588 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
Unit Size:
100 pairs
Product Code. 17899662
Brand: Ansell M2079
Only null left
Add to basket
Add to basket
Synthetic, powder-free examination gloves that are ideal for frequent glove changes.
  • This non-sterile, synthetic glove is made from copolymer materials (ethylene vinyl acetate film) and is a safe and effective alternative to natural rubber latex.
  • li>Packaged on backing paper for easy donning
  • Cost effective synthetic alternative
  • Protects from latex Type I allergy
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 455 Part 1, EN 455 Part 3, EN 455 Part 1, EN 455 Part 3, ISO 10993-10:2002, ISO 13485
Certifications/Compliance (Europe) EN 455 Part 1, EN 455 Part 3, EN 455 Part 1, EN 455 Part 3, ISO 10993-10:2002, ISO 13485
Chemotherapeutic Rated No
Accelerator Free No
Applications Healthcare
AQL 1.5
Color Clear
Disposable Single-use
Exterior Finish No Texture
Finger Thickness (English) 1.2 mil
Finger Thickness (Metric) 0.03 mm
Latex Free Yes
Material EVA
Palm Thickness (English) 1.2 mil
Palm Thickness (Metric) 0.03 mm
Powdered Powder Free
Product Type Examination Gloves
Size Medium
Thickness (English) 1.2 mil
Thickness (Metric) 0.03 mm
FDA 510 (k) No
Model ETHIPARAT Non-Sterile
Length (English) 10.9 in.
Length (Metric) 278 mm