missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 associated protein II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-11491-0.05MG
This item is not returnable.
View return policy
Description
ETS1 associated protein II Polyclonal specifically detects ETS1 associated protein II in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| ETS1 associated protein II | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:500 | |
| AD022, dJ30M3.3, EAP2, EAPII, EC 3.1.4.-, ETS1-associated protein 2, ETS1-associated protein II, MGC111021,5′-Tyr-DNA phosphodiesterase, MGC9099,5′-tyrosyl-DNA phosphodiesterase, TRAF and TNF receptor associated protein, TTRAPTRAF and TNF receptor-associated protein, Tyr-DNA phosphodiesterase 2, tyrosyl-DNA phosphodiesterase 2 | |
| Rabbit | |
| 0.05 mg | |
| 51567 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide | |
| TDP2 | |
| The synthetic peptide taken from human Tyrosyl-DNAphosphodiesterase (TDP2) protein corresponding to amino acids spanning 335-356. The peptide was conserved in several other species. The selected peptide was post-synthetically modified to achieve highest antigenicity before coupling to carrier protein using hetero bifunctional cross linker for immunogen preparation | |
| Primary | |
| The antibody labels a strong band of TDP2 of 45kDa in TDP12 samples and in several cell lines. The antibody also labels a diffuse band of around 95 and 115kDa, the identity of these bands is not known but may represent TDP2 splice variants. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction