missing translation for 'onlineSavingsMsg'
Learn More

ETS1 associated protein II Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 251.00 - € 745.00

Specifications

Antigen ETS1 associated protein II
Dilution Western Blot 1:500, ELISA 1:500
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18098108
View Documents
Novus Biologicals
NBP2-11491-0.025MG
0.025 mg
€ 251.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18067968
View Documents
Novus Biologicals
NBP2-11491-0.05MG
0.05 mg
€ 437.00
0.05mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18027169
View Documents
Novus Biologicals
NBP2-11491
0.1 mg
€ 745.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ETS1 associated protein II Polyclonal specifically detects ETS1 associated protein II in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

ETS1 associated protein II
Western Blot, ELISA
Unconjugated
Rat, Human, Mouse
TDP2
IgG
The antibody labels a strong band of TDP2 of 45kDa in TDP12 samples and in several cell lines. The antibody also labels a diffuse band of around 95 and 115kDa, the identity of these bands is not known but may represent TDP2 splice variants.
Western Blot 1:500, ELISA 1:500
Polyclonal
Rabbit
51567
The synthetic peptide taken from human Tyrosyl-DNAphosphodiesterase (TDP2) protein corresponding to amino acids spanning 335-356. The peptide was conserved in several other species. The selected peptide was post-synthetically modified to achieve highest antigenicity before coupling to carrier protein using hetero bifunctional cross linker for immunogen preparation
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.