Learn More
EVOS™ EVOS™ FL Auto Master Plate for Large Vessels
Product Code. 15389651
Click to view available options
Quantity:
1 plate
Unit Size:
Each
Product Code. 15389651
Brand: EVOS™ AMEPVH027
Designed for use with EVOS™ FL Auto Imaging System
  • Allows user to utilize different types of vessels for imaging of cells grown in large cell culture flasks and dishes
  • Suitable for T150 and larger tissue culture flasks
  • Has easy-to-change plates and holders available for this instrument platform, for optimal precision in sample alignment
TRUSTED_SUSTAINABILITY
Specifications
Type Master Plate
Product Line EVOS
For Use With (Equipment) EVOS™ FL Auto Imaging System
Holds Large Tissue Culture Flask, T150 Vessel
Quantity 1 plate