missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Evsco™ PVDF Two-Way Elliptic Valve

Use elliptic valves for liquid, gas and vacuum applications

Brand:  Cole-Parmer™ 06470-47

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11825362

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Easy to disassemble: Simply pull out the handle and push your stem through the bottom, allowing you to clean and maintain valves in-line
  • Four-position valves are clearly marked with flow patterns to ensure correct handle position
  • Choose from FDA-grade PP or PVDF bodies
  • All valves have O-rings made of Viton™, 316 SS handles, threaded connections and nonrestrictive orifices
  • Rated up to 150 psi at 77°F and 230°F (110°C) at 50 psi (liquids)
Specifications

Specifications

2-Way Manual Elliptical Valve
0.5 in.
PVDF, Viton
1/2 in. NPT(F)
White
NPT(F) Connection
12.7 mm
1/2 in. NPT(F)
1/2 in. NPT(F)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers