missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14805-1-AP-150UL
This item is not returnable.
View return policy
Description
TRMT1 is a tRNA(m(2)(2)G(26))dimethyltransferaseSpecifications
| EXOSC2 | |
| Polyclonal | |
| Unconjugated | |
| EXOSC2 | |
| EXOSC2, exosome component 2, hRrp4p, p7, RRP4, Rrp4p | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 227715, 23404, 366017 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13868, Q8VBV3 | |
| Exosc2 | |
| EXOSC2 Fusion Protein Ag6525 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction