missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 87-950 (ex EXTRA)
Extra protection for heavy duty applications.
Brand: Ansell™ 87-950_9.5-10
Additional Details : Weight : 0.10000kg
Description
- 50% thicker than standard flocklined gloves.
- Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
- 100% natural rubber with no fillers. Excellent resistance to mechanical hazards plus increased chemical protection.
- Chlorinated. For better grip and increased chemical resistance.
- 100% cotton flocklining. Makes a heavy glove softer and more comfortable to wear. Helps absorb perspiration.
Specifications
Rubber Latex | |
Natural Rubber Latex | |
No | |
10 | |
0.75 mm | |
Straight | |
144Pack | |
87-950 |
EN388: 2003, EN374:2003, EN388:2016, EN ISO 374-1:2016, EN ISO 374-5:2016 | |
Natural Rubber Latex | |
Immersion Gloves | |
87-950 | |
Black | |
Cotton Flocking | |
Many ketones, salts, detergents, alcohols, alkalis and fats |