missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves

Provides for heavy duty handling where sensitivity is also required. Ansell Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves offer extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.

Brand:  Ansell™ 87-955_8.5-9

 View more versions of this product

Product Code. 15222879

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 50% thicker than standard flocklined gloves
  • Chlorinated for better grip and increased chemical resistance
  • 100% natural rubber with no fillers for excellent resistance to mechanical hazards
  • 100% cotton flocking for softness, comfort and increased absorption
  • Ideal for chemical processing and preparation and maintenance of plant and heavy equipment
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rubber Latex
Natural Rubber Latex
Immersion Gloves
87-955
Orange
Cotton Flocking
Many ketones, salts, detergents, alcohols, alkalis and fats
Natural Rubber Latex
No
9
0.75 mm
Straight
144Pack
87-955
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers