Learn More
Fisherbrand™ Eyepiece Pair for Magnum Series, 12.5x
Product Code. 10142544 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
12.5X
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power unitSize
10142544 12.5X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10142544
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for use with Magnum Series microscopes. Fisherbrand™ Eyepiece Pairs are ergonomically designed for everyday use.
Magnum series microscopes have a robust T-shaped base, designed for maximum stability, with modern ergonomic features to ensure a comfortable, prolonged viewing position.
  • The trinocular versions allow connection to cameras whilst the planachromatic objectives provide true colour reproduction excluding spherical aberration
  • The phase contrast models allow an efficient view of thin or translucent specimens where staining is difficult
  • Capable of Kohler illumination, accessories are also available for darkfield and polarization enabling a wide range of applications
  • Head is inclined at 30° with interpupillary distance adjustment (Siedentopf type) of 55 to 75mm
  • The quintuple reversed nosepiece revolves with click stop
  • Includes: blue and green filter, 1 x spare bulb, 2 x spare fuses, immersion oil 10mL, soft rubber eyecups, dust cover and instruction manual
  • Images may illustrate the microscope with Coolpix digital camera accessory: please note this is an optional extra and is NOT supplied as standard. For details, please contact Customer Service.
Specifications
Type Widefield Eyepiece Pair
For Use With (Application) Magnum Series microscopes.
Magnification Power 12.5X