missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAAH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35192-100ul
This item is not returnable.
View return policy
Description
FAAH Polyclonal antibody specifically detects FAAH in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| FAAH | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Anandamide amidohydrolase 1, EC 3.5.1, EC 3.5.1.99, EC 3.5.1.n2, FAAH1, FAAH-1, fatty acid amide hydrolase, fatty-acid amide hydrolase 1, MGC102823, MGC138146, Oleamide hydrolase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-579 of human FAAH (NP_001432.2).,, Sequence:, EIEVYRKTVIAQWRALDLDVVLTPMLAPALDLNAPGRATGAVSYTMLYNCLDFPAGVVPVTTVTAEDEAQMEHYRGYFGDIWDKMLQKGMKKSVGLPVAVQCVALPWQEELCLRFMREVERLMTPEKQSS | |
| 100 μL | |
| Lipid and Metabolism | |
| 2166 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction