missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAAH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | FAAH |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227315
|
Novus Biologicals
NBP3-35192-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230893
|
Novus Biologicals
NBP3-35192-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAAH Polyclonal antibody specifically detects FAAH in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| FAAH | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 2166 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Anandamide amidohydrolase 1, EC 3.5.1, EC 3.5.1.99, EC 3.5.1.n2, FAAH1, FAAH-1, fatty acid amide hydrolase, fatty-acid amide hydrolase 1, MGC102823, MGC138146, Oleamide hydrolase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-579 of human FAAH (NP_001432.2).,, Sequence:, EIEVYRKTVIAQWRALDLDVVLTPMLAPALDLNAPGRATGAVSYTMLYNCLDFPAGVVPVTTVTAEDEAQMEHYRGYFGDIWDKMLQKGMKKSVGLPVAVQCVALPWQEELCLRFMREVERLMTPEKQSS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto