missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP8/M-FABP/Myelin P2 Protein Antibody (3H3F4D5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06650-50ul
This item is not returnable.
View return policy
Description
FABP8/M-FABP/Myelin P2 Protein Monoclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISA
Specifications
| FABP8/M-FABP/Myelin P2 Protein | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3H3F4D5 | |
| ELISA 1:1000-1:2000 | |
| M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PMP2 (Accession#: NP_002668.1; Ser 2-Val 132). | |
| 50 μg | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 5375 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction