missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM65B Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04522-100UL
This item is not returnable.
View return policy
Description
FAM65B Polyclonal specifically detects FAM65B in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FAM65B | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9750 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| C6orf32, DIFF48DIFF40, family with sequence similarity 65, member B, PL48KIAA0386chromosome 6 open reading frame 32 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human FAM65B (NP_056948.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction