missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FAM70B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30755-100ul
This item is not returnable.
View return policy
Description
FAM70B Polyclonal antibody specifically detects FAM70B in Human samples. It is validated for Western Blot
Specifications
| FAM70B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| family with sequence similarity 70, member B, FLJ57938, MGC20579, RP11-199F6.1 | |
| Produced in rabbits immunized with E. coli-derived Human FAM70B fragment | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS, pH 7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 348013 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction