missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FAM70B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 195.00 - € 396.00
Specifications
| Antigen | FAM70B |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30042907
|
Novus Biologicals™
NBP3-30755-20ul |
20 μL |
€ 195.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30042723
|
Novus Biologicals™
NBP3-30755-100ul |
100 μL |
€ 396.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM70B Polyclonal antibody specifically detects FAM70B in Human samples. It is validated for Western BlotSpecifications
| FAM70B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| family with sequence similarity 70, member B, FLJ57938, MGC20579, RP11-199F6.1 | |
| Produced in rabbits immunized with E. coli-derived Human FAM70B fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.0 | |
| 348013 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title