Learn More
FAM81A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-33295
 View more versions of this product
Product Code. 18776903
  • € 549.00 / 0.10mL
Description
FAM81A Polyclonal specifically detects FAM81A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
family with sequence similarity 81, member A, hypothetical protein LOC145773, MGC26690
Affinity Purified
IgG
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50