missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM81A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 280.00 - € 549.00
Specifications
| Antigen | FAM81A |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18420362
|
Novus Biologicals
NBP2-33295-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776903
|
Novus Biologicals
NBP2-33295 |
€ 549.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
FAM81A Polyclonal specifically detects FAM81A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FAM81A | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| family with sequence similarity 81, member A, hypothetical protein LOC145773, MGC26690 | |
| FAM81A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8TBF8 | |
| 145773 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LFLEEHIRNITAIVKQLNRDIEVLQEQIRARDNISYGTNSALKTLEMRQLSGLGDLRGRVARCDASIARLSAEHKTTYEGLQHLNKEQQAAKLILETKIKDAEGQISQLLNRVDLSISEQSTKLKMSHRDSNHQLQLLDT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title