Learn More
FASN Rabbit, anti-Human, CoraLite Plus 488, Clone: 1L9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech CL488-81079-20UL
 View more versions of this product
Product Code. 18018195
  • € 145.00 / 20µL
Description
The FASN antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human FASN. This antibody recognizes human, mouse, rat antigen. The FASN antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
FASN
1L9
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Immunofluorescence, Flow Cytometry
1000 μg/mL
FAS, FASN, fatty acid synthase, OA 519, SDR27X1
20 μL
2194
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.