Learn More
FASN Rabbit, anti-Human, CoraLite Plus 488, Clone: 1L9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech CL488-81079-20UL
Description
The FASN antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human FASN. This antibody recognizes human, mouse, rat antigen. The FASN antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™ plus 488.
Specifications
| FASN | |
| 1L9 | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Immunofluorescence, Flow Cytometry | |
| 1000 μg/mL | |
| FAS, FASN, fatty acid synthase, OA 519, SDR27X1 | |
| 20 μL | |
| 2194 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |