Learn More
FASN Rabbit, anti-Human, CoraLite Plus 488, Clone: 1L9, Proteintech
Rabbit Recombinant Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | FASN |
|---|---|
| Clone | 1L9 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074295
|
Proteintech
CL488-81079-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18018195
|
Proteintech
CL488-81079-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FASN antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human FASN. This antibody recognizes human, mouse, rat antigen. The FASN antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™ plus 488.
Specifications
| FASN | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 2194 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1L9 | |
| Immunofluorescence, Flow Cytometry | |
| Liquid | |
| Human, Mouse, Rat | |
| FAS, FASN, fatty acid synthase, OA 519, SDR27X1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.