missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXW8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
€ 300.00 - € 572.00
Specifications
| Antigen | FBXW8 |
|---|---|
| Concentration | 0.2mg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18470691
|
Novus Biologicals
NBP1-84715-25ul |
25ul |
€ 300.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18290488
|
Novus Biologicals
NBP1-84715 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXW8 Polyclonal specifically detects FBXW8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FBXW8 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 26259 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KLIFQECRAKEHMLQTNWKNRKGAVSELEHVPDTVLCDVHSHDGVVIAGYTSGDVRVWDTRTWDYVAPFLESEDEEDEPGMQPNVSFVRINSSLAVAA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.2mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| F-box and WD repeat domain containing 8, F-box only protein 29F-box/WD repeat-containing protein 8, FBW8F-box and WD-40 domain-containing protein 8, FBX29MGC33534, FBXO29, FBXW6FBW6F-box and WD-40 domain protein 8 | |
| FBXW8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title