missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDX1 Antibody (SR1148), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 446.25
Specifications
| Antigen | FDX1 |
|---|---|
| Clone | SR1148 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671105
|
Novus Biologicals
NBP3-22548-100ul |
100 μg |
€ 472.00 € 446.25 / 100µL Save € 25.75 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652634
|
Novus Biologicals
NBP3-22548-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FDX1 Monoclonal antibody specifically detects FDX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| FDX1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2230 | |
| IgG | |
| Affinity purified |
| SR1148 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Adrenal ferredoxin, adrenodoxin, mitochondrial, ADXhepatoredoxin, FDX, ferredoxin 1, ferredoxin-1, Hepatoredoxin, LOH11CR1D, mitochondrial adrenodoxin | |
| A synthesized peptide derived from human FDX1 (Uniprot #: P10109) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title