missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEN-1 Antibody (S01-3I5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19863-50ul
This item is not returnable.
View return policy
Description
FEN-1 Monoclonal antibody specifically detects FEN-1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| FEN-1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S01-3I5 | |
| Unconjugated | |
| EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2 | |
| A synthetic peptide of human FEN-1 (Uniprot # P39748) | |
| 50 μg | |
| Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases | |
| 2237 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction