missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Heavy Chain Antibody (962610) - Azide and BSA Free, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB9354
This item is not returnable.
View return policy
Description
Ferritin Heavy Chain Monoclonal specifically detects Ferritin Heavy Chain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Ferritin Heavy Chain | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 5 μg/mL, Immunocytochemistry 1 μg/mL | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 962610 | |
| Unconjugated | |
| PBS with No Preservative | |
| FTH1 | |
| The immunogen is raised against Ferritin Heavy Chain. | |
| 100 μg | |
| Lipid and Metabolism | |
| 2495 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction