missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Heavy Chain Antibody (962610) - Azide and BSA Free, R&D Systems™
Mouse Monoclonal Antibody
€ 494.00
Specifications
| Antigen | Ferritin Heavy Chain |
|---|---|
| Clone | 962610 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 5 μg/mL, Immunocytochemistry 1 μg/mL |
| Applications | Western Blot |
Description
Ferritin Heavy Chain Monoclonal specifically detects Ferritin Heavy Chain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Ferritin Heavy Chain | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| PBS with No Preservative | |
| 2495 | |
| The immunogen is raised against Ferritin Heavy Chain. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 962610 | |
| Western Blot 5 μg/mL, Immunocytochemistry 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| FTH1 | |
| IgG2a | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title