missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetal Hemoglobin Antibody (SR1113), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22184-100ul
This item is not returnable.
View return policy
Description
Fetal Hemoglobin Monoclonal antibody specifically detects Fetal Hemoglobin in Human samples. It is validated for Western Blot
Specifications
| Fetal Hemoglobin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3047 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1113 | |
| Western Blot 1:500-1:2000 | |
| abnormal hemoglobin, A-gamma globin, gamma A hemoglobin, gamma globin, gamma-1-globin, gamma-2-globin, gamma-globin chain, G-gamma globin Paulinia, Hb F Agamma, hb F Ggamma, HBGA, HBGR, HBG-T1, HBG-T2, Hemoglobin gamma-1 chain, hemoglobin gamma-2 chain, Hemoglobin gamma-A chain, hemoglobin gamma-G chain, hemoglobin subunit gamma-1, hemoglobin subunit gamma-2, hemoglobin, gamma A, hemoglobin, gamma, regulator of, HSGGL1, methemoglobin, PRO2979 | |
| A synthesized peptide derived from human Fetal Hemoglobin (Uniprot #: P69891, P69892) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction