missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetal Hemoglobin Antibody (SR1113), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 352.80 - € 456.75
Specifications
| Antigen | Fetal Hemoglobin |
|---|---|
| Clone | SR1113 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621054
|
Novus Biologicals
NBP3-22184-100ul |
100 μg |
€ 483.00 € 456.75 / 100µL Save € 26.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676554
|
Novus Biologicals
NBP3-22184-50ul |
50 μg |
€ 373.00 € 352.80 / 50µL Save € 20.20 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fetal Hemoglobin Monoclonal antibody specifically detects Fetal Hemoglobin in Human samples. It is validated for Western BlotSpecifications
| Fetal Hemoglobin | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3047 | |
| IgG | |
| Affinity purified |
| SR1113 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| abnormal hemoglobin, A-gamma globin, gamma A hemoglobin, gamma globin, gamma-1-globin, gamma-2-globin, gamma-globin chain, G-gamma globin Paulinia, Hb F Agamma, hb F Ggamma, HBGA, HBGR, HBG-T1, HBG-T2, Hemoglobin gamma-1 chain, hemoglobin gamma-2 chain, Hemoglobin gamma-A chain, hemoglobin gamma-G chain, hemoglobin subunit gamma-1, hemoglobin subunit gamma-2, hemoglobin, gamma A, hemoglobin, gamma, regulator of, HSGGL1, methemoglobin, PRO2979 | |
| A synthesized peptide derived from human Fetal Hemoglobin (Uniprot #: P69891, P69892) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title