missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57305-25ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FGF-12 Polyclonal specifically detects FGF-12 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spécification
| FGF-12 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| FGF-12, FGF12B, FHF-1, FHF1Fibroblast growth factor homologous factor 1, fibroblast growth factor 12, fibroblast growth factor 12B, fibroblast growth factor FGF-12b, Myocyte-activating factor | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| FGF12 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EPSLHEIGEKQGRSRKSSGTPTMNGGKVVNQDS | |
| 25 μL | |
| Angiogenesis | |
| 2257 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu