missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41227
This item is not returnable.
View return policy
Description
FGFR3 Polyclonal specifically detects FGFR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FGFR3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FGFR3 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human FGFR3. The immunogen is located within amino acids 630 - 680 of FGFR3. Amino Acid Squence: RDVHNLDYYKKTTNGRLPV | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least three isoforms of FGFR3 are known to exist; this antibody will detect all three. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 - 2 ug/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Rabbit | |
| 92, 98 kDa | |
| 0.1 mg | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction