missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 189.00 - € 551.00
Specifications
| Antigen | FGFR3 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647646
|
Novus Biologicals
NBP2-41227-0.025mg |
0.025 mg |
€ 189.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18098583
|
Novus Biologicals
NBP2-41227 |
0.1 mg |
€ 551.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR3 Polyclonal specifically detects FGFR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FGFR3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| FGFR3 | |
| IgG | |
| Affinity Purified | |
| 92, 98 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Protein Kinase | |
| PBS with 0.02% Sodium Azide | |
| 2261 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human FGFR3. The immunogen is located within amino acids 630 - 680 of FGFR3. Amino Acid Squence: RDVHNLDYYKKTTNGRLPV | |
| Primary | |
| At least three isoforms of FGFR3 are known to exist; this antibody will detect all three. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title