missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (CL3730), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61633
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Fibronectin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| FN1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HEEICTTNEGVMYRIGDQWDKQHDMGHMMRCTCVGNGRGEWTCIAYSQLRDQCIVDDITYNVNDTFHKRHEEGHMLNCTCFGQGRGRWKCDPVDQCQDSETGTFYQIGDSWEKYVHGVRYQCYCYGRGIG | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| CL3730 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:1000 - 1:2500 | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Mouse | |
| Protein A purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| 2335 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction