missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (CL3730), Novus Biologicals™
Mouse Monoclonal Antibody
€ 277.00 - € 511.00
Specifications
| Antigen | Fibronectin |
|---|---|
| Clone | CL3730 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240464
|
Novus Biologicals
NBP2-61633 |
100 μL |
€ 511.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632669
|
Novus Biologicals
NBP2-61633-25ul |
25 μL |
€ 277.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Fibronectin | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| 2335 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HEEICTTNEGVMYRIGDQWDKQHDMGHMMRCTCVGNGRGEWTCIAYSQLRDQCIVDDITYNVNDTFHKRHEEGHMLNCTCFGQGRGRWKCDPVDQCQDSETGTFYQIGDSWEKYVHGVRYQCYCYGRGIG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL3730 | |
| Monoclonal | |
| Purified | |
| Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| FN1 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title