missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 5/DANCE Antibody (SR1879), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21755-50ul
This item is not returnable.
View return policy
Description
Fibulin 5/DANCE Monoclonal antibody specifically detects Fibulin 5/DANCE in Human, Mouse samples. It is validated for Western Blot
Specifications
| Fibulin 5/DANCE | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10516 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1879 | |
| Western Blot 1:500-1:2000 | |
| ARMD3fibulin-5, Dance, DANCEFIBL-5, Developmental arteries and neural crest EGF-like protein, EVEC, fibulin 5, UP50FLJ90059, Urine p50 protein | |
| A synthesized peptide derived from human Fibulin 5/DANCE (Uniprot #: Q9UBX5) | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction