missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 5/DANCE Antibody (SR1879), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Fibulin 5/DANCE |
|---|---|
| Clone | SR1879 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670884
|
Novus Biologicals
NBP3-21755-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632725
|
Novus Biologicals
NBP3-21755-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibulin 5/DANCE Monoclonal antibody specifically detects Fibulin 5/DANCE in Human, Mouse samples. It is validated for Western BlotSpecifications
| Fibulin 5/DANCE | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 10516 | |
| IgG | |
| Affinity purified |
| SR1879 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| ARMD3fibulin-5, Dance, DANCEFIBL-5, Developmental arteries and neural crest EGF-like protein, EVEC, fibulin 5, UP50FLJ90059, Urine p50 protein | |
| A synthesized peptide derived from human Fibulin 5/DANCE (Uniprot #: Q9UBX5) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title