Learn More
Filaggrin (Keratinocyte Differentiation Marker) Mouse, Clone: FLG/1563, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio 2312-MSM3-P0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15929997
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Filaggrin (Keratinocyte Differentiation Marker)
Cytoplasmic
FLG/1563
Unconjugated
Mouse
26-45kDa (Processed); 350kDa (Profilaggrin)
Protein A/G purified from Bioreactor concentrate.
RUO
2312
IgG2b κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human Filaggrin protein fragment (apprx. aa 998-1104)
U-251-MG cells or Skin
20 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.