Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Shop All Fisherbrand Products
Click to view available options
Holds:
0.2 mL PCR Plate
0.5 mL Tubes
1.5 mL Tubes
15 mL Tubes
2 mL Tubes
384-well Plate
5 mL Eppendorf Tubes
50 mL Tubes
Microplate
Well Count:
1
4
9
24
30
96
384
Product Type:
0.2mL PCR Plate Block with Lid
384-well Plate Block with Lid
Microplate Thermal Block with lid
Tube Block
Unit Size:
Each
Specifications
Specifications
| Holds | Microplate |
| Product Type | Microplate Thermal Block with lid |
| Depth (English) | 0.9 in. |
| Depth (Metric) | 2.3 cm |
| Dimensions (L x W) | 12.9 x 8.6 cm (5.06 x 3.4 in.) |
| For Use With (Application) | Heating/shaking |
| For Use With (Equipment) | Shake Touch Shakers |
| Well Count | 1 |