Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Shop All Fisherbrand Products
Click to view available options
Holds:
0.2 mL PCR Plate
0.5 mL Tubes
1.5 mL Tubes
15 mL Tubes
2 mL Tubes
384-well Plate
5 mL Eppendorf Tubes
50 mL Tubes
Microplate
Well Count:
1
4
9
24
30
96
384
Product Type:
0.2mL PCR Plate Block with Lid
384-well Plate Block with Lid
Microplate Thermal Block with lid
Tube Block
Unit Size:
Each
Specifications
Specifications
| Holds | 384-well Plate |
| Product Type | 384-well Plate Block with Lid |
| Depth (English) | 0.32 in. |
| Depth (Metric) | 0.81 cm |
| Diameter (English) | 0.16 in. |
| Diameter (Metric) | 0.4 cm |
| Dimensions (L x W x H) | 4.7 x 6.4 x 3.0 in. (11.9 x 16.3 x 7.6 cm) |
| For Use With (Application) | Heating/Shaking |
| For Use With (Equipment) | Shake Touch Shakers |
| Well Count | 384 |