Learn More
Fisherbrand™ Isotemp™ Shake Touch Shakers Accessory Blocks
Shop All Fisherbrand Products
Click to view available options
Holds:
0.2 mL PCR Plate
0.5 mL Tubes
1.5 mL Tubes
15 mL Tubes
2 mL Tubes
384-well Plate
5 mL Eppendorf Tubes
50 mL Tubes
Microplate
Well Count:
1
4
9
24
30
96
384
Product Type:
0.2mL PCR Plate Block with Lid
384-well Plate Block with Lid
Microplate Thermal Block with lid
Tube Block
Unit Size:
Each
Specifications
Specifications
| Holds | 5 mL Eppendorf Tubes |
| Tube Capacity | 5 mL |
| Product Type | Tube Block |
| Depth (English) | 1.93 in. |
| Depth (Metric) | 4.9 cm |
| Diameter (English) | 0.660 in. |
| Diameter (Metric) | 1.68 cm |
| Dimensions (L x W x H) | 4.2 x 5.7 x 5.0 in. (10.7 x 14.5 x 12.7 cm) |
| For Use With (Application) | Heating/shaking |
| For Use With (Equipment) | Shake Touch Shakers |
| Show More |