Learn More
Fisherbrand™ Labtool II™ and Mini-Labtool™
Shop All Fisherbrand ProductsDescription
Includes
Labtool II includes pliers/gripper, wire stripper, wire cutter, ruler, small/large knife, serrated knife, punch, flat head and Phillips screwdrivers, bottle opener, needle, saw, and serrated stripper
Mini-Labtool includes pliers/gripper, wire stripper, wire cutter, knife, flat head and Phillips screwdrivers, scissors, file, saw, bottle and can openers, and key chain
Open bottles; cut stoppers, tubing, and wires; and much more. Pocket-size, Fisherbrand™ multi-use tools are ideal for hundreds of lab needs.
- Stainless steel with aluminum handles
Order Info
Labtool II, closed length: 3.5 in. (9cm); weight, 7 oz. (200g).
Mini-Labtool, closed length, 2.5 in. (6.3cm); weight, 3 oz. (85g).
Specifications
Specifications
| Product Type | Mini Labtool |
| Length (English) | 2.5 in. |
| Length (Metric) | 6.3 cm |
| Material | Stainless Steel |
| Size | Pocket-size |
| For Use With (Application) | Opener for chemical bottle, saw to reduce cork stopper, knife to cut tubing |
| Includes | Pliers/gripper, wire stripper, wire cutter, knife, flat head and Phillips™ screwdrivers, scissors, file, saw, bottle and can openers and key chain |
| Weight (English) | 0.18 lb. |
| Weight (Metric) | 0.085 kg |