Learn More
Fisherbrand™ PrepSep™ Large-Volume SPE Columns
Versatile design provides rapid flowthrough using syringe, vacuum box, or robot technology
POA
Product Code | Brand | Column Capacity (Metric) | Sorbent | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity (Metric) | Sorbent | Price | Quantity & Availability | ||||
10194954
|
Fisherbrand™
|
12 mL | Empty |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Sorbent packings separate and retain sample analytes from their matrices; retrieve analytes by eluting with an appropriate solvent
- Packing type is clearly marked on upper rim of each column for easy identification
- Economically disposable; inexpensive enough to use once then throw away, eliminating cross-contamination and cleanups
- Large-reservoir columns feature 12 to 60mL reservoirs that accept larger sample volumes; also feature a conical shape for easy filling
Specifications
SPE |