Learn More
Fisherbrand™ PrepSep™ Large-Volume SPE Columns
Versatile design provides rapid flowthrough using syringe, vacuum box, or robot technology
POA
Specifications
Column Type SPE
View More Specs
Products 1
Product Code Brand Column Capacity (Metric) Sorbent Price Quantity & Availability  
Product Code Brand Column Capacity (Metric) Sorbent Price Quantity & Availability  
10194954
View Documents Promotion Details
Fisherbrand™
12 mL Empty This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Sorbent packings separate and retain sample analytes from their matrices; retrieve analytes by eluting with an appropriate solvent
  • Packing type is clearly marked on upper rim of each column for easy identification
  • Economically disposable; inexpensive enough to use once then throw away, eliminating cross-contamination and cleanups
  • Large-reservoir columns feature 12 to 60mL reservoirs that accept larger sample volumes; also feature a conical shape for easy filling
Specifications
Specifications
SPE