missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIZZ1/RELM alpha Antibody (RM0313-9A39) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12440-0.025MG
This item is not returnable.
View return policy
Description
FIZZ1/RELM alpha Monoclonal specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FIZZ1/RELM alpha | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9EP95 | |
| RETNLA | |
| Mouse | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| RM0313-9A39 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| 1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 57262 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction