missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIZZ1/RELM alpha Antibody (RM0313-9A39) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 206.00 - € 507.00
Specifications
| Antigen | FIZZ1/RELM alpha |
|---|---|
| Clone | RM0313-9A39 |
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18018936
|
Novus Biologicals
NBP2-12440-0.025MG |
0.025 mg |
€ 206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18045667
|
Novus Biologicals
NBP2-12440 |
0.1 mg | |||||||
Description
FIZZ1/RELM alpha Monoclonal specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FIZZ1/RELM alpha | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9EP95 | |
| 57262 | |
| IgG2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| RM0313-9A39 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2 | |
| RETNLA | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title