Learn More
Honeywell™ FLEXIDYN Light Task Plus 4 Gloves
Product Code. p-7159571
Click to view available options
Size:
7, Small
8, Medium
9, Large
10, X-Large
11, 2X-Large
Unit Size:
1 pair
Product Code. 15876805
Brand: Honeywell™ NFG17X/7S
Only null left
Add to basket
Add to basket
Honeywell™ FLEXIDYN Light Task Plus 4 Gloves for handling sharp parts in a dry, oily or greasy environment.
  • Nylon and glass fiber knitted glove 3/4 Bi polymer (PU and Nitrile) coating knitted wrist
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The Glass fiber protected by polyamide thread provides very good properties against cuts
  • The 3/4 Bi polymer coating provides outstanding abrasion and perforation resistance and protects well the knuckle
  • Its Dark color enables work in a slightly soiled environment
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC PPE Category 2, EC Certificate Number 0075/797/162/06/18/1075
Description 10 gauge
Color Gray, Blue
Material Nylon, Glass Fiber
Size 7, Small
Length (Metric) 25 cm