missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flotillin 1 Recombinant Rabbit Monoclonal Antibody (23GB845), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554077
This item is not returnable.
View return policy
Description
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
Specifications
| Flotillin 1 | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75955 | |
| FLOT1 | |
| A synthetic peptide of human Flotillin 1 (400-427AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB845 | |
| Unconjugated | |
| FLOT1 | |
| DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10211 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction