missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flotillin 1 Recombinant Rabbit Monoclonal Antibody (23GB845), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | Flotillin 1 |
|---|---|
| Clone | 23GB845 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088097
![]() |
Invitrogen™
MA554076 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088742
|
Invitrogen™
MA554077 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Flotillin 1 Recombinant Monoclonal antibody specifically detects Flotillin 1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Flotillin 1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75955 | |
| 10211 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB845 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2 | |
| A synthetic peptide of human Flotillin 1 (400-427AA). | |
| Primary | |
| -20°C | |
| FLOT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title