Learn More
FMC63 scFv, Rabbit anti-Mouse, Polyclonal Antibody, ACROBiosystems
Rabbit Polyclonal Antibody
Brand:  ACROBiosystems FM3-S93-100ug
 View more versions of this product
Product Code. 17266210
  • € 358.00 / 100µg
Description
Description
Recognizes FMC63 scFv specifically, (recommended for ADA assay) (MALS verified)
Specifications
Specifications
FMC63
Polyclonal
Lyophilized from 0.22μm filtered solution in PBS, pH7.4. Normally trehalose isadded as protectant before lyophilization.
100 μg
Primary
Mouse
Anti-idiotypic Antibodies
ELISA
Unconjugated
Rabbit
RUO
Recognizes FMC63 scFv specifically
-20°C
Powder