Learn More
FMC63 scFv, Rabbit anti-Mouse, Polyclonal Antibody, ACROBiosystems
Rabbit Polyclonal Antibody
Brand: ACROBiosystems FM3-S93-100ug
Description
Recognizes FMC63 scFv specifically, (recommended for ADA assay) (MALS verified)Specifications
| FMC63 | |
| Polyclonal | |
| Lyophilized from 0.22μm filtered solution in PBS, pH7.4. Normally trehalose isadded as protectant before lyophilization. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Anti-idiotypic Antibodies |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| Recognizes FMC63 scFv specifically | |
| -20°C | |
| Powder |