missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMC63 scFv, Rabbit anti-Mouse, Polyclonal Antibody, ACROBiosystems
Rabbit Polyclonal Antibody
€ 358.00 - € 2580.00
Specifications
| Antigen | FMC63 |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Powder |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17266210
|
ACROBiosystems
FM3-S93-100ug |
100 μg |
€ 358.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17276210
|
ACROBiosystems
FM3-S93-1mg |
1 mg |
€ 2580.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recognizes FMC63 scFv specifically, (recommended for ADA assay) (MALS verified)Specifications
| FMC63 | |
| Polyclonal | |
| Powder | |
| RUO | |
| Lyophilized from 0.22μm filtered solution in PBS, pH7.4. Normally trehalose isadded as protectant before lyophilization. | |
| -20°C | |
| Anti-idiotypic Antibodies |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Primary | |
| Recognizes FMC63 scFv specifically |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title