missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR2, Rat anti-Mouse, Clone: CL10, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-81079-0.2MG
This item is not returnable.
View return policy
Description
FOLR2 Monoclonal specifically detects FOLR2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FOLR2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
| Rat | |
| Protein A purified | |
| RUO | |
| 2350 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry | |
| CL10 | |
| Unconjugated | |
| PBS | |
| FOLR2 | |
| FRbeta -transfected RBL2H3 cells. | |
| 0.2 mg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction